Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Barium titanium oxide, polymeric precursor, (Oxide ^= 35 wt%), Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
Description
Barium titanium oxide is used in piezoelectric devices. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Viscous Liquid |
Quantity | 2 g |
UN Number | UN1993 |
Solubility Information | Soluble in alcohols |
Concentration or Composition (by Analyte or Components) | Oxide ≈35 wt% |
Chemical Name or Material | Barium titanium oxide, Polymeric precursor |
Safety and Handling
Hazard Category | H302+H332 |
Hazard Statement | GHS H Statement H226-H302-H332 Flammable liquid and vapor. Harmful if swallowed. Harmful if inhaled. |
Precautionary Statement | P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
TSCA | Yes |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction