Alfa Aesar™ Barium titanium oxide, polymeric precursor, (Oxide » 35 wt%)

Used in piezoelectric devices

Manufacturer: Alfa Aesar™ 3975504

Quantity 2g

 View more versions of this product

Catalog No. AA3975504

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12047-27-7
    MDL Number MFCD00003447
    UN Number UN1993
    Chemical Name or Material Barium titanium oxide, Polymeric precursor

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P261-P280-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H226-H302-H332
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful if inhaled.
    MDL Number MFCD00003447
    CAS 12047-27-7
    UN Number UN1993
    Solubility Information Soluble in alcohols
    Physical Form Viscous Liquid
    Quantity 2g
    Health Hazard 1 Warning
    Concentration Oxide ≈35 wt%
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material Barium titanium oxide, Polymeric precursor

    GHS H Statement
    H226-H302-H332
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P210-P261-P280-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit