Barium trifluoroacetate hydrate, White crystalline, Alfa Aesar™

Manufacturer: Alfa Aesar 44596-09

Quantity 10g

 View more versions of this product

Catalog No. AA4459609

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Physical Form Crystals
    Molecular Formula Ba(OOCCF3)2·xH2O
    Formula Weight 363.37
    MDL Number MFCD00137143
    Chemical Name or Material Barium trifluoroacetate hydrate

    Specifications

    Physical Form Crystals
    Formula Weight 363.37
    Quantity 10g
    Color White
    DOT Information DOT Class: 6.1, Packaging Group: III
    Anhydrous or Hydrate Hydrate
    Molecular Formula Ba(OOCCF3)2·xH2O
    MDL Number MFCD00137143
    Health Hazard 1 H302-H332: Harmful if swallowed. Harmful if inhaled.
    Chemical Name or Material Barium trifluoroacetate hydrate

    H302-H332: Harmful if swallowed. Harmful if inhaled.

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit