Alfa Aesar™ Barium trifluoromethanesulfonate

Manufacturer: Alfa Aesar™ 3857104

Quantity 2g

 View more versions of this product

Catalog No. AA3857104

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2794-60-7
    Formula Weight 435.48
    MDL Number MFCD00143644
    UN Number UN1564
    Beilstein 3729346
    Synonym Barium triflate
    Chemical Name or Material Barium trifluoromethanesulfonate

    Specifications

    Formula Weight 435.48
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00143644
    UN Number UN1564
    Health Hazard 1 Warning
    Sensitivity Hygroscopic
    Beilstein 3729346
    Melting Point >300°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    Physical Form Powder
    CAS 2794-60-7
    Synonym Barium triflate
    Quantity 2g
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    Chemical Name or Material Barium trifluoromethanesulfonate

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit