Alfa Aesar™ Barium trifluoromethanesulfonate

Manufacturer: Alfa Aesar™ 3857109

Quantity 10g

 View more versions of this product

Catalog No. AA3857109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2794-60-7
    Formula Weight 435.48
    MDL Number MFCD00143644
    UN Number UN1564
    Beilstein 3729346
    Synonym Barium triflate
    Chemical Name or Material Barium trifluoromethanesulfonate

    Specifications

    Formula Weight 435.48
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    Physical Form Powder
    Beilstein 3729346
    Health Hazard 1 Warning
    Quantity 10g
    Sensitivity Hygroscopic
    Synonym Barium triflate
    Melting Point >300°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
    TSCA TSCA
    MDL Number MFCD00143644
    CAS 2794-60-7
    UN Number UN1564
    Chemical Name or Material Barium trifluoromethanesulfonate

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit