Alfa Aesar™ Barium zirconium isopropoxide, 10% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles

Precursor of barium zirconium oxide using Sol-Gel method

Manufacturer: Alfa Aesar™ 42302AP

Quantity 500mL

 View more versions of this product

Catalog No. AA42302AP

  Check Availability
Add to Cart

    Chemical Identifiers

    Formula Weight 583.08
    MDL Number MFCD00798551
    UN Number UN3274
    Chemical Name or Material Barium zirconium isopropoxide

    Specifications

    Formula Weight 583.08
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00798551
    Sensitivity Moisture Sensitive
    UN Number UN3274
    Quantity 500mL
    Health Hazard 2 GHS H Statement
    H225-H302-H332-H336
    Highly flammable liquid and vapor.
    Harmful if swallowed.
    Harmful if inhaled.
    May cause drowsiness or dizziness.
    Packaging ChemSeal™ bottles
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
    Physical Form Liquid
    Concentration 10% w/v in isopropanol
    Storage Note 1
    • Packaged under Argon
    Chemical Name or Material Barium zirconium isopropoxide

    GHS H Statement
    H225-H302-H332-H336
    Highly flammable liquid and vapor.
    Harmful if swallowed.
    Harmful if inhaled.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P280-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit