Learn More
Thermo Scientific™ Barnstead™ 1/2 Size B-Pure™ Filters Available on GSA/VA Contract for Federal Government customers only.
Pre-treat your feed water with Thermo Scientific™ Barnstead™ 1/2 Size B-Pure™ Filters, available in varying pore sizes.
Manufacturer:  Thermo Scientific™ 18011
 View more versions of this product
Catalog No. 09-034-153
Add to cart
Description
Description
Pre-treat your feed water with Thermo Scientific™ Barnstead™ 1/2 Size B-Pure™ Filters. In addition to pre-treating feed water, the filters can act as a final filter to ensure consistently pure water. Compatible with B-Pure 1/2 size filter holders, these filters are available in varying pore sizes to meet all of your pre-filtration needs.
  • Large surface area for long life
  • Flow rates of up to 2L/min.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1/2 Size B-Pure Filter Holders
10µm Pre-filter for Economy Filter Holder
6.5cm
25.4cm
Up to 2L/min
2.5 in.
10 in.
10µm