Learn More
Thermo Scientific™ Barnstead™ 1/2 Size B-Pure™ Pressure Gauges
Monitor pressure drop across any of the 10 in. filter cartridges used in the 1/2 size B-Pure™ filter holder with a Thermo Scientific™ Barnstead™ 1/2 Size B-Pure™ Pressure Gauge.
$161.40
Specifications
For Use With (Equipment) Any B-Pure System
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
09-034-93
View Documents
Thermo Scientific™
D0780
Pressure Gauge, Single Each for $161.40
Add to cart
Description
Description
Monitor pressure drop across any of the 10 in. filter cartridges used in the B-Pure™ 1/2 size filter holder with a Thermo Scientific™ Barnstead™ 1/2 Size B-Pure™ Pressure Gauge.
  • Attach to any B-Pure system 0 to 160psi pressure gauge, allowing for monitoring inlet and outlet water pressure.
  • Monitor pressure drop across any of the 10 in. filter cartridges used in the 1/2 size B-Pure filter holder.
  • Pressure drop of greater than 10 psi indicates need for filter change.
Specifications
Specifications
Any B-Pure System