Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Barnstead™ Classic Stills Available on GSA/VA Contract for Federal Government customers only.

Remove inorganic solids, organics, bacteria and pyrogens from your water with Thermo Scientific™ Barnstead™ Classic Stills.

Manufacturer:  Thermo Scientific™ A1013C

 View more versions of this product

Catalog No. 09-026-5


  Check Availability
Add to cart

Specifications

Description

Remove inorganic solids, organics, bacteria and pyrogens from your water through the distillation process using Thermo Scientific™ Barnstead™ Classic Stills. Choose from bench or floor mounted models. Available in five capacities to meet your production needs, stills are constructed of copper and bronze with a pure tin coating. The inert nature of tin prevents leaching of contaminants into water. Portable stills allow for quick set-up, while floor model electric stills improve safety with over-temperature protection. Steam stills feature optional automatic controls for unattended operation.

  • Double-walled boiler and preheating of feed water conserves electricity.
  • Vented condenser allows for stripping of gaseous impurities.
  • Unique de-concentrator removes scale forming impurities from the boiler.
  • Unique Q-Baffle ensures high-quality pyrogen-free water by stripping contaminant laden water droplets from steam.
  • Still includes space-saving horizontal condenser.
  • Metal construction withstands years of use.
  • Inert pure tin pathways assure product water quality.

Portable Stills:

  • Require no permanent plumbing or electrical connections allowing for quick setup
  • Include low water cutoff for safety protection
  • Bench mounted

Floor Model Electric Still:

  • Comes standard with floor mounting stand
  • Improves safety with new overtemperature protection system
  • Allows for easier installation with new plug-and-play hardware
  • Processes capacities from 1 to 10 gal./hr.
  • Includes low water cutoff that protects heating elements when water supply is interrupted
  • Allows for unattended operation with optional fully automatic controls
  • Stops still when storage tank is full, starts still when water level falls below preset level
  • Drains still boiler at selected two-, four-, eight- and 16-hour intervals to reduce scale formation

Steam Still:

  • Utilizes available in-house steam as heat source
  • Optional fully automatic controls allow for unattended operation
  • Stops still when storage tank is full, starts still when water level falls below preset level
  • Drains still boiler at selected two-, four-, eight- and 16-hour intervals to reduce scale formation
  • Steam pressure: 35 to 50psi
  • Boiler horsepower: 3.5
  • Steam flowrate: 90 lb./hr. (41kg/hr.)

Ordering Information:

Automated control systems are available for all stills: G2100 (for 1 gal./hr., 2 gal./hr. models), G2110 (5 gal./hr.), G2125 (10 gal./hr.) and G2010 (10 gal./hr.). Order separately.

This product is not available in all regions.

Includes:

Floor stand (exept for Portable Model A1007).

Requires:

Storage reservoir, UV light assembly and Ventgard Air Filter (order separately)

Recommended for:

Buffer/reagent prep, glassware washing, dishwasher feed

Description & Specifications

Specifications

11 x 23 x 68 in. (28 x 58 x 173cm)
Electric
208V
17A
68 in.
23 in.
16 gal./hr.
11 in.
2 gal./hr.
UL, cUL
208V 50/60Hz
2 gal/hr (204V) Floor Electric with stand
50/60Hz
3
173cm
58cm
61L/hr.
28cm
7.6L/hr.
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit