Learn More
Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables Available on GSA/VA Contract for Federal Government customers only.
Maintain the performance of your Thermo Scientific™ Barnstead™ Classic Still with replacement consumables such as UV lamps and air filters.
$131.00 - $361.50
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
09-028-45 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
04141
Replacement UV Lamp Each for $131.00
Add to cart
09-050-116
View Documents
Thermo Scientific™
H3120
replacement filter for H3111 Each for $361.50
Add to cart
Description
Description
Maintain the performance of your still with Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables, including UV lamp and Thermo Scientific™ Ventgard™ filter element (the consumable portion of the Ventgard filter assembly). Prevent air from entering the reservoir via the condenser atmospheric vent and protect stored water against airborne particulates, organics and CO2 with the Ventgard air filter. 
Maintain the performance of your still with Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables, including UV lamp and Thermo Scientific™ Ventgard™ filter element (the consumable portion of the Ventgard filter assembly). Prevent air from entering the reservoir via the condenser atmospheric vent and protect stored water against airborne particulates, organics and CO2 with the Ventgard air filter. 
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.