Learn More
Thermo Scientific™ Barnstead™ Classic Still Storage Accessories Available on GSA/VA Contract for Federal Government customers only.
Complete your distillation system with Thermo Scientific™ Barnstead™ Classic Still Storage Accessories.
Manufacturer:  Thermo Scientific™ H4005
 View more versions of this product
Catalog No. 09-028-42
Add to cart
Description
Description
Thermo Scientific™ Barnstead™ Classic Still Storage Accessories complement your current system to create a more complete system. Accessories include mounting options, UV light assembly, Thermo Scientific™ Ventgard™ air filter and water seal. Choose either a floor stand or wall bracket as needed. Maintain sterility in the reservoir with the UV light assembly, which includes housing, on/off switch, 6 ft. cord, lamp and ballast. Protect stored water against airborne particulates, organics and CO2 with Ventgard™ air filter and water seal. Prevent air from entering the reservoir via the condenser atmospheric vent.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Classic reservoirs
UV Light Assembly
UV Light Assembly
Maintains water sterility in tank. Includes house, on/off switch, 6 ft. cord, lamp and ballast