Learn More
Thermo Scientific™ Barnstead™ Easypure™ Cartridges and Filters Available on GSA/VA Contract for Federal Government customers only.
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with replacement cartridges and filters.
$98.20 - $713.00
Products 7
Catalog Number Mfr. No. For Use With Description Price Quantity & Availability  
Catalog Number Mfr. No. For Use With Description Price Quantity & Availability  
09-034-165 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
D502124
Easypure II UV and Easypure II UV/UF Cartridge pack; deionized feed Each for $594.00
Add to cart
09-034-166 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
D502125
Easypure II UV and Easypure II UV/UF Cartridge pack; RO or distilled feed Each for $594.00
Add to cart
09-034-167
View Documents
Thermo Scientific™
D502126
Easypure II and Easypure UF Cartridge pack; deionized feed Each for $579.00
Add to cart
09-034-168 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
D502127
Easypure II and Easypure UF Cartridge pack; RO or distilled feed Each for $545.50
Add to cart
13-687-217 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
FL1192X1
Easypure UF and Easypure II UV/UF Ultrafilter Each for $713.00
Add to cart
09-034-17 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
D50230
Each for $175.30
Add to cart
09-050-66
View Documents
Thermo Scientific™
D50245
Each for $98.20
Add to cart
Description
Description
Extend the life of your Thermo Scientific™ Barnstead™ Easypure™ II or Easypure™ RF water system with Barnstead™ Easypure™ Cartridges and Filters. Choose the cartridge pack according to your feed water and system.
D502126:
  • Includes: Organic/Colloid; two Thermo Scientific™ Ultrapure™ Ion Exchange
  • Type 1 filter
D502124:
  • Includes: Organic/Colloid, Ultrapure Ion Exchange, Organic Free Organic Removal
  • Ultralow filter with Organic Free Cartridge
D502127:
  • Includes: Organic/Ion Exchange, two Ultrapure Ion Exchange
  • Type 1 filter
D502125:
  • Includes: 09-034-18 Organic/Ion Exchange, 09-050-46 Ultrapure Ion Exchange, 09-034-16 Organic Free Organic Removal
  • Ultralow filter with Organic Free Cartridge
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.