Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Barnstead™ FI-Streem™ Glass Stills Pretreatment Kits

Treat samples with the Thermo Scientific™ Barnstead™ FI-Streem™ Glass Stills Pretreatment Kit when feed water quality does not match system requirements.

Manufacturer:  Thermo Scientific™ A56285

 View more versions of this product

Catalog No. 09-047-87

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Treat samples with the Thermo Scientific™ Barnstead™ FI-Streem™ Glass Stills Pretreatment Kit when feed water quality does not match system requirements.

  • Utilizes disposable cartridges
  • D x W x H: 8.28 x 11 x 23.5 in. (21 x 27.9 x 59.7cm)
Specifications

Specifications

Barnstead™ FI-Streem&tgrade; Glass Still
Pretreatment Kit, 120V
Pretreatment Kit, 120V
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit