Learn More
Thermo Scientific™ Barnstead™ FI-Streem™ Glass Stills Pretreatment Kits
Treat samples with the Thermo Scientific™ Barnstead™ FI-Streem™ Glass Stills Pretreatment Kit when feed water quality does not match system requirements.
Specifications
For Use With (Equipment) Barnstead™ FI-Streem™ Glass Still
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
09-047-87
View Documents
Thermo Scientific™
A56285
Pretreatment Kit, 120V This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Treat samples with the Thermo Scientific™ Barnstead™ FI-Streem™ Glass Stills Pretreatment Kit when feed water quality does not match system requirements.
  • Utilizes disposable cartridges
  • D x W x H: 8.28 x 11 x 23.5in. (21 x 27.9 x 59.7cm)
Specifications
Specifications
Barnstead™ FI-Streem™ Glass Still