Learn More
Thermo Scientific™ Barnstead™ GenPure™ Pro Water Purification System UV PROMO Available on GSA/VA Contract for Federal Government customers only.
Suitable for the most demanding laboratory applications, the Barnstead™ GenPure™ Pro water purification system delivers ultrapure 18.2 megohm-cm water with consistent quality.
Manufacturer:  Thermo Scientific™ 50131950INS
 View more versions of this product
Catalog No. 10-451-215PM
Add to cart
Description
Description
Dispensing for a variety of tasks:
  • New flexible dispenser offers a radius of 24 in. (60cm) from the system for filling larger vessels or glassware washing.
Integrated feed water monitoring:
  • An additional measuring cell monitors feed water conductivity. Any exceeding of the limiting value is immediately displayed to maximize cartridge life.
UV Photo-oxidation 185/254nm:
  • High performance UV assembly reliably reduces the content of microorganisms and their metabolites.
  • UV oxidation also reduces organic compounds in the water to ultra-low levels.
Ready-to-use:
  • Feed water pressure switch, wall bracket, UV lamp, ultrapure cartridge, sterile filter, are all included in one part number.
  • Leak detection is standard and will alert user to leaks.
Measurement of conductivity/resistivity:
  • Conductivity cells are carefully calibrated prior to each measurement via built-in reference resistance with cell constants at 0.01 cm-1.
  • Temperature measurements are made by a platinum chip sensor with ±0.1°C accuracy.
Exact dosing:
  • High precision POM draw-off valve offers dispensing with sterile filter.
  • System offers electronic dosing for fully automatic volume control of 0.01-65.0L with <2% accuracy.
GLP-compliant documentation
  • Real-time clock and code-protected operating system prevents unauthorized changes to system settings.
  • System includes RS-232 interface with adjustable send interval for safe data transfer of all measured data, faults, date and time to a PC computer or log printer.
  • Digital microprocessor control automatically monitors and stores faults from the last four weeks.
  • USP-compliant conductivity measurement with temperature compensation can be switched on or off.
Tilting Control Panels for Easy Viewing:
  • Ergonomically designed controller tilts for ease-of-use and better visibility
  • Panel offers illuminated four-line alpha-numeric display.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
18.2MΩ·cm
Up to 2L/min
<0.001 EU/mL
<1
24.2 in.
13 in.
2 to 6 bar
50/60Hz
18.6 in.
GenPure Pro ultrapure water system with UV-photo-oxidation and ultrafiltration membrane
0.1kW
0.055µS/cm
1 to 5ppb
<0.1 CFU/mL
3/4 in. NPT
615mm
330mm
90-240V
2° - 35°C
472mm
Barnstead GenPure Pro UV/UF
90-240V 50/60Hz
Documents
Accessories