Learn More
Thermo Scientific™ Barnstead™ Hose Nipple Cartridges Available on GSA/VA Contract for Federal Government customers only.
Purify water up to 75 L/hr using Thermo Scientific™ Barnstead™ Hose Nipple Cartridges for routine lab work including pretreatment.
Manufacturer:  Thermo Scientific™ D8902
 View more versions of this product
Catalog No. 09-035-25
Add to cart
Description
Description
Purify water volumes up to 75L/hr using cost-efficient Thermo Scientific™ Barnstead™ Hose Nipple Cartridges. Ideal for softening, deionization, organic and chlorine removal applications, these cartridges are for use with the simple cartridge holder that easily mounts to the wall. Designed for non-pressure applications, these cartridges include 3/8 in. barb hose connections on each end. See when resin is exhausted in specific cartridges by color change indicator. Constructed of 100% virgin polypropylene cartridge, the system is customizable and an ideal option for routine laboratory work.
  • Ideal for small volume applications
  • Same superior quality resins used in B-Pure cartridges
  • Constructed of 100% virgin polypropylene cartridge
  • Includes 3/8 in. barb hose connections on each end D8822, D8950, and D8951 cartridges include tapered straight nipple for 3/8 in. ID tubing
  • Color change indicator that identifies when resin is exhausted in specific cartridges
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Ultrapure Mixed-Bed Ion Exchange
915 grains
Up to 75L/hr.
Barnstead™ Water Purification System
Ultrapure Mixed-Bed Ion Exchange
Yes
Full Size Ultrapure