Learn More
Thermo Scientific™ Barnstead™ Hose Nipple Cartridges
GSA_VA
Purify water up to 75 L/hr using these hose nipple cartridges, ideal for routine lab work including pretreatment.
$73.10 - $237.00
Specifications
Flow Rate Up to 75 L/hr.
View More Specs
Products 11
Catalog Number Mfr. No. Description Resin Type Price Quantity & Availability  
Catalog Number Mfr. No. Description Resin Type Price Quantity & Availability  
09-050-264
View Documents
Thermo Scientific™
D8822
1/2 Size Mixed-Bed with Oxygen Removal Ultrapure Mixed Bed with Strong Anion Exchange
Each for $205.00
Add to cart
09-035-20
GSA_VA
Required Accessories View Documents
Thermo Scientific™
D8901
Full Size High Capacity Two Bed Ion Exchange
Each for $237.00
Edge
Add to cart
09-035-25
GSA_VA
View Documents
Thermo Scientific™
D8902
Full Size Ultrapure Ultrapure Mixed Bed Ion Exchange
Each for $237.00
Edge
Add to cart
09-035-22
GSA_VA
Required Accessories View Documents
Thermo Scientific™
D8904
Full Size Organic Removal Activated Carbon
Each for $178.80
Add to cart
09-034-12
View Documents
Thermo Scientific™
D8905
Hose nipple cartridge, Full Size Cation Removal Cation Exchange
Each for $176.70
Add to cart
09-034-14
Required Accessories View Documents
Thermo Scientific™
D8911
Full Size Ultrapure Ultrapure Ion Exchange
Each for $229.50
Add to cart
09-035-32
GSA_VA
Required Accessories View Documents
Thermo Scientific™
D8921
Full Size Pretreatment and Scale Eliminator Mixed Bed and Strong Cation Exchange, Activated Charcoal
Each for $219.50
Add to cart
09-035-30
GSA_VA
Required Accessories View Documents
Thermo Scientific™
D8922
Full Size Ultrapure with Organic Removal Mixed Bed Ion Exchange with Activated Carbon
Each for $235.50
Add to cart
09-035-28
Required Accessories View Documents
Thermo Scientific™
D8950
2/3 Size High Capacity Two Bed Ion Exchange with Strong Cation Exchange
Each for $183.90
Add to cart
09-035-029
View Documents
Thermo Scientific™
D8951
2/3 Size Mixed-Bed and Organic Removal Mixed Bed Ion Exchange with Activated Carbon
Each for $222.00
Add to cart
09-035-50
View Documents
Thermo Scientific™
D8900
Hose Nipple Cartridge Holder
Each for $73.10
Add to cart
Description
Description
Purify water volumes up to 75L/hr using cost-efficient Thermo Scientific™ Barnstead™ Hose Nipple Cartridges. Ideal for softening, deionization, and organic and chlorine removal applications, these cartridges are for use with the simple cartridge holder that easily mounts to the wall. Designed for non-pressure applications, these cartridges include 3/8″ barb hose connections on each end. They provide the ability to see when resin is exhausted in specific cartridges by a color change indicator. Constructed of 100% virgin polypropylene cartridge, the system is customizable and an ideal option for routine laboratory work.
  • Ideal for small volume applications
  • Same superior quality resins used in B-Pure cartridges
  • Constructed of 100% virgin polypropylene cartridge
  • Includes 3/8 in. barb hose connections on each end
  • D8822, D8950, and D8951 cartridges include tapered straight nipple for 3/8 in. ID tubing
  • Color change indicator that identifies when resin is exhausted in specific cartridges
Specifications
Specifications
Up to 75 L/hr.