Learn More
Thermo Scientific™ Barnstead™ MegaPure™ Bottles
Safely store distilled water using plastic or glass Thermo Scientific? Barnstead? MegaPure? Bottles compatible with Barnstead Mega-Pure Glass Stills.
Manufacturer:  Thermo Scientific™ 410535
 View more versions of this product
Catalog No. 09-083
  Check Availability
Add to cart
Specifications
Description
Safely store distilled water using plastic or glass Thermo Scientific™ Barnstead™ MegaPure™ Bottles compatible with Barnstead Mega-Pure Glass Stills. Water bottles for this distillation system are available in varying capacities.
Description & Specifications
Specifications
2.43 gal., 9L
MP-1 and MP-3A stills
Glass
9L
MP-1 and MP-3A Stills
9L (2.43 gal.), Glass bottle with stopcock
2.43 gal.