Learn More
Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems Ultrafilters
GSA_VA
Lengthen the life of your water purification system with these replacement ultrafilters, which are automatically flushed.
$717.00 - $1611.00
Specifications
Type Water Purification Systems Ultrafilter
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
10-451-110
GSA_VA
View Documents
Thermo Scientific™
50133980
GenPure, GenPure Pro, GenPure xCAD
Each for $1,611.00
Add to cart
10-451-111
GSA_VA
View Documents
Thermo Scientific™
50133981
MicroPure, Smart2Pure (UF and UF/UV units only)
Each for $717.00
Edge
Add to cart
Description
Description
Lengthen the life of your water purification system with replacement Thermo Scientific™ Barnstead™ Point-of-Use Water Purification Systems Ultrafilters. For use with Barnstead point-of-use water purification systems, these internal ultrafilters are flushed automatically to allow for a long two-year lifetime.
Use with the following Barnstead point-of-use water purification systems:
Part 50133980:
  • GenPure™†
  • GenPure™ Pro†
  • GenPure™ xCAD†
†UF, UF/UV and UF/UV-TOC units only
Part 50133981:
  • MicroPure™‡
  • Smart2 Pure™‡
‡UF and UF/UV units only
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Water Purification Systems Ultrafilter