Thermo Scientific™ Barnstead™ TII Type 2 Water System

This item is no longer available and has been replaced by the next generation of Barnstead Water Purification Systems.

Manufacturer: Thermo Scientific™ D14031DEL

 View more versions of this product

Catalog No. 09-036-10

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Particles >0.2µM/mL <1 μM/mL
    Bacterial Content <1CFU/mL
    Resistivity 15MΩ·cm at 25°C
    TOC Monitor with UV Intensity Monitoring No
    Feed Water Monitoring No
    Feedwater Pressure 30 to 100psig (2 to 6.9 bar)
    Height (English) 19.5 in.
    Height (Metric) 49.5cm
    Width (English) 13.5 in.
    Width (Metric) 34.3cm
    Voltage 100/240VAC
    Hertz 50/60Hz
    Inlet Water Temperature (Metric) 4° to 40°C
    Conductivity 0.067 to 0.1 µS/cm
    Bacterial retention 99%
    Depth (English) 18.5 in.
    Depth (Metric) 47cm
    Description Discontinued; replacement product is 50132131
    Purified Water Flowrate Field 1L/min.
    Feedwater Source tap
    RO Flow Rate 12L/hr.
    Certifications/Compliance CE, CSA
    Electrical Requirements 100/240VAC 50/60Hz
    Total Organic Carbon 1 to 5ppb

    This item is no longer available and has been replaced by the next generation of Barnstead Water Purification Systems. The recommended replacement for this product is the Thermo Scientific™ Barnstead™ Pacific™ TII water purification system.

    Produce Type 2 pure water that exceeds ASTM and CLSI-CLRW standards with the smart and flexible Thermo Scientific&trade; Barnstead&trade; TII Type 2 Water System. This system is ideal for all applications requiring pure water such as the preparation of microbiological media and buffers, glassware washing and as feed water for Type 1 ultrapure water systems.

    Features:
    • Green/yellow/red indicator lights simplify operation with detailed status on alert information.
    • System is available in 12L and 24L/hr. production rates with 30L or 60L storage tanks for a variety of application needs.
    • System monitors water at all points including feedwater, RO water, final product water.
    • System measures resistivity and temperature.
    • Removable display can be mounted up to 10 ft. (3m) from unit for added flexibility.
    • Space-saving design: unit can be wall-mounted or placed on benchtop UV lamp status can be monitored on display to ensure bacterial-free, low-organic water.
    • Cartridge disconnects easily for simple, fast replacement.
    • Fluorinated PE reservoir eliminates leaching of organic materials and protects against contamination.
    • Direct dispensing offers options of volumetric (0.25 to 60L) or timed dispense.
    Smart, Easy to Use, Low-Maintenance System:
    • Multi-step purification combines pretreatment, reverse osmosis, deionization, UV oxidation and filtration in one.
    • System easily dispenses from unit or reservoir.
    • Exclusive flow pattern and unique water storage and recirculation system continuously maintain stored water.
    • Smart controls monitor critical system data and automate operation, ensuring maximum water quality.
    • Sophisticated multifunctional display provides real-time information on water quality, system performance and diagnostics.
    • Intuitive multifunction main menu has push-button controls to monitor critical system data.
    • System allows easy access to alarm information, setup data and real-time operational readings such as water purity, percent RO rejection, cartridge status and reservoir levels.

    Ordering Information:

    Optional accessories include remote dispensers, printer, cutoff float, interlock cable, storage reservoirs, bottles and replacement consumables. These products are available separately. For details, contact your sales representative.

    Includes:

    • Wall bracket
    • RO membrane(s)
    • UV lamp

    Requires:

    • Start-Up Kit (D502138)
    • Storage reservoir

    Recommended for:

    • Media prep
    • Buffer preps
    • Glassware washing
    • Equipment feed
    • Type 1 Water system feed

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit