Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Tough polyethylene tape comes in a dispenser box and has a repeating warning message.
Order Info
Thickness: 2mil. Dimensions: 1000 ft.L × 3 in. W (304.8m × 7.6cm).
Specifications
Yellow | |
1000 ft. | |
HDPE | |
1000 ft. x 3 in. (304.8 m x 7.62 cm) | |
0.002 mil | |
304.8 m |
Dispenses quickly and easily to effectively designate a hazardous area | |
3 in. | |
Barricade Tape | |
Caution | |
7.62 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title