Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barricade Tape
SCI_ED

Specifications

Color Yellow
Dimensions (L x W) 1000 ft. x 3 in. (304.8 m x 7.62 cm)
Material HDPE
Type Barricade Tape
For Use With (Application) Dispenses quickly and easily to effectively designate a hazardous area
View More Specs

Products 1
Catalog Number Mfr. No. Displayed Text Price Quantity & Availability  
Catalog Number Mfr. No. Displayed Text Price Quantity & Availability  
S85375
SCI_ED
View Documents
Honeywell Safety Products
CT3YE1/EMD
Caution This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Tough polyethylene tape comes in a dispenser box and has a repeating warning message.

Order Info

Thickness: 2mil. Dimensions: 1000 ft.L × 3 in. W (304.8m × 7.6cm).

Specifications

Specifications

Yellow
HDPE
Dispenses quickly and easily to effectively designate a hazardous area
304.8 m
3 in.
1000 ft. x 3 in. (304.8 m x 7.62 cm)
Barricade Tape
1000 ft.
0.002 mil
7.62 cm
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit