Learn More
Barrier™ Cone Facemasks
Manufacturer:  MOLNLYCKE 4233102
 View more versions of this product
Catalog No. 17-989-42
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
ASTM F2101, ASTM F1215
Soft breathable manmade Copolymer with hypoallergenic innerfacing and non-glass design
Copolymer
Soft arch, pleated
Blue
Tie Strings
97% (BFE), 98% (PFE)