Barrier™ Ear Loop Facemasks
Range of styles and levels of protection
Manufacturer: MOLNLYCKE 4229101
 View more versions of this product
Catalog No. 17-989-33
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications/Compliance ASTM F2101, ASTM F1215
    Filter Efficiency >97% (BFE), 98% (PFE)
    Color Blue
    Construction Highly fluid-repellent 4-ply Meltblown laminate material with special non-woven Fibers between each of the layers ensures breathability, adjustable nosepiece
    Resistance Fluid
    Style Pleated
    Pore Size 0.6μ
    Includes Soft Elastic Earloops
    Material Meltblown
  • Description & Specifications
Related Products
Product Content Feedback