Learn More
Molnlycke Barrier Tie Band Anti-Fog Face Mask
Specifications
Certifications/Compliance | ASTM F2101, ASTM F1215 |
---|---|
Color | Green |
Construction | Non-glass design with Plastic film across the nosebridge to prevent eyeglasses from fogging |
Pore Size | 0.3μ |
Style | Pleated |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
17-989-39
|
Molnlycke
42381 |
Without tie-strings |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. Due to product restrictions, please Sign In to purchase or view availability for this product. |
N/A |
Description
Highly breathable pleated style provides a cool, comfortable fit. Pleated style maximizes coverage area. Choose from a basic facemask or the antifog mask, which features a special plastic film across the nosebridge to prevent eyeglasses from fogging. Both masks have soft tie strings and offer a BFE greater than 95% for particle sizes down to 0.6 micron. Green. In packs of 50.
Order Info
Green. In packs of 50.
Specifications
ASTM F2101, ASTM F1215 | |
Non-glass design with Plastic film across the nosebridge to prevent eyeglasses from fogging | |
Pleated |
Green | |
0.3μ |