Learn More
Barrier™ Tie Band Anti-Fog Face Mask
Manufacturer:  MOLNLYCKE 42391
 View more versions of this product
Catalog No. 17-989-40
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
ASTM F2101, ASTM F1215
Non-glass design with Plastic film across the nosebridge to prevent eyeglasses from fogging
Basic
Yes
0.3μ
Green
With tie-strings
95% (BFE)
Pleated