Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BarX1 Mouse anti-Human, Clone: BARX1/2759, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | BarX1 |
---|---|
Clone | BARX1/2759 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122878
|
Novus Biologicals
NBP30770220UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
BarX1 Monoclonal specifically detects BarX1 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
BarX1 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
BarH-like homeobox 1, BARX homeobox 1, homeobox protein BarH-like 1 | |
Recombinant fragment of human BarX1 protein (around aa 155-254) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
BARX1/2759 | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
56033 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title