Learn More
Eisco™ Basalt Specimen
SCI_ED
Basalt is formed quickly from an iron-rich magma and exhibits a dark color and small crystal size.
Supplier:  Eisco™ ESNG0041PK10
 View more versions of this product
Catalog No. S26589
Explore available promotions
Add to Cart
Description
Description
  • Aphanitic volcanic rock forms from effusive eruptions, such as shield volcanoes of Hawaii, or deep ocean floor
  • For use in introduction to igneous rock classification and identification
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Students can recreate cooling history of this sample based on texture and abundance of phenocrysts
  • Example of aphanitic igneous rock
Specifications
Specifications
Igneous Rock, Obsidian
10/Pk.
2 to 3 cm
0.2 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.