Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Base for Polystyrene Plant Tissue Container

Bottom part holders for tissue culture containers.

$237.00

Specifications

Material Polystyrene
Color Clear
Description Base for plant tissue container (accessory)
View More Specs

Products 1
Catalog Number Mfr. No. Sterility For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Sterility For Use With (Application) Price Quantity & Availability  
07-000-962 View Documents Greiner Bio-One
960177
Sterile Cultivation of Drosophila melanogaster Case of 315 for $237.00
Add to cart
 
Description

Description

  • Bottom part base
  • For use with Polystyrene plant tissue culture container (Part No: 960177)

Service Plan

No

Warranty and Services

No

Specifications

Specifications

Polystyrene
Clear
Base for plant tissue container (accessory)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit