Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ BASIC™ Universal Absorbent Rolls
BASIC Universal Sorbents are economical, general purpose sorbents to clean up oils, coolants, solvents, and water-based fluids.
Supplier: Brady™ BRU152
Description
- Universal absorbency designed to offer an economical, general purpose solution to cleaning
- Sorbents absorb up to 20 times their weight in fluids
- Continuous rolls are ideal for covering larger areas
- Perforations every 30 in. make it easy to select smaller sizes when needed
- Use against spills in assembly lines, printing, equipment or any general manufacturing areas
- Economical single-ply construction
Specifications
14.5 gl/gs | |
Perforated every 30 in.; split roll | |
Polypropylene | |
2 Rolls | |
150 ft. x 15 in. (45.7 m x 38 cm) |
Gray | |
38 gal. | |
Chemical resistant sorbent for water, oil and solvent base liquids | |
Heavyweight |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction