Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Basic Digital Stereo Zoom Microscope

Basic Entry Level Stereo Zoom Microscope for Macro-Size Specimens is ideal for educational and laboratory use

Catalog No. 03-000-039


Add to cart

Description

Description

Optical System

  • 10X WF eyepiece
  • 1X to 4X objective
  • Continuous zoom control
  • Zoom ratio: 1:4

Viewing Head

  • Trinocular viewing head with 45° incline
  • Interpupillary adjustment of 52mm to 75mm
  • Diopter Adjustment
  • 0.5x C-mount lens
  • 3MP, USB, Color camera

Focusing

  • Coarse focus adjustment
  • 40mm height adjustment range on focusing mechanism
  • 70mm working distance

Stand

  • Low profile plain stage with clips
  • Solid Stand
  • Compact design

Illumination

  • Eco-friendly and energy efficient LED illumination
  • LED ring in transmitted light, provides homogeneous light distribution
  • LED incident Illumination
  • Membrane switch controls
Specifications

Specifications

Limited lifetime
WF10X
1x to 4x
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit