Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Basic Fuchsin, Electrophoresis Reagent
Supplier: Thermo Scientific™ J6649414
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
632-99-5 | |
Crystalling powder | |
268° to 270°C (dec.) | |
Plastic bottle | |
C.I. 42510,Fuchsin basic |
Ambient temperatures | |
337.86 | |
C20H20ClN3 | |
25g |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction