Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Basic Fuchsin, Electrophoresis Reagent

$18.14 - $178.40

Specifications

Melting Point 268° to 270°C (dec.)
Format Crystalling powder
Synonym C.I. 42510,Fuchsin basic
Molecular Formula C20H20ClN3
CAS 632-99-5
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ6649406 View Documents Alfa Aesar™
J6649406
5g Each for $18.14
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ6649414 View Documents Alfa Aesar™
J6649414
25g Each for $62.30
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ6649422 View Documents Alfa Aesar™
J6649422
100g Each for $178.40
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

268° to 270°C (dec.)
C.I. 42510,Fuchsin basic
632-99-5
337.86
Crystalling powder
C20H20ClN3
Plastic bottle
Safety and Handling

Safety and Handling

EINECSNumber : 211-189-6

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit