Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3B Scientific™ Basic Life Support Simulator, BASICBilly+

Shop All 3B Scientific Products


Click to view available options
Color:
Dark Skin
Light Skin
Description
Solid and Compact Manikin for Effective Training:
- Made of easy to clean durable, latex free material
- Hygienic, with easy to change disposable airways
- Realistic anatomical landmarks to learn correct hand position for chest compression
- Mouth-to-mouth, mouth-to-nose or BVM resuscitation can be practiced
- Offers head-tilt functionality to open the airway or simulate obstructed airway
- Visible chest rise when the lungs are ventilated
- Force required for effective chest compressions has been carefully studied to offer a realistic training approach
- Operation requires (2) AA batteries, not included
The CPR Apps Provide Feedback For:
- Compression (depth, recoil, and rate)
- Breath (volume)
- Hands-off time in real-time
- Feedback and evaluation meet the standard of the latest guidelines of the American Heart Association (AHA) and the European Resuscitation Council (ERC)
Overview of the CPR App Features:
- Offers overall scoring and intuitive point of improvement during CPR
- Direct (real-time) and summative CPR feedback for students
- Instructor view can monitor up to 6 manikins at a time
- Saved student results for review
- Export or share students result/certificate as a PDF file
- Instructor can use CPR analysis for detailed debriefing
- Easily switch from conventional CPR training to Compression Only CPR settings
- Stable connection is provided by a reliable Bluetooth connection
- Mobile devices used to access the apps are not included in the delivery scope
- An Upgrade Kit (1022626) is available to equip previously purchased BASICBilly manikins with the AHA required direct feedback functionalities
Specifications
Specifications
| Product Type | AHA Compliant, BLS Manikin |
| For Use With (Application) | Train, Monitor, Analyze, and Debrief CPR Performances |
| Color | Light Skin |
| Includes | BASICBilly BLS Manikin (1), Adult Lung Bags (6), Adult Cardiac Massage Spring (1), Face Mask with Insert and Lung Bag Holder (1), Carrying Bag with Training Mat (1), BASICBilly+ Upgrade Kit (1), Add-On Kit with Breath Module Sensor (1), ColorID Stickers (1), Installation Manual (1) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction