Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Basic Nuclear Lab Stations
Fully integrated system is capable of performing many basic nuclear demonstrations.
Manufacturer: Spectrum Techniques Inc PO210S.1
Includes: The Nuclear Lab Station
Supplied complete with a set of three radioisotope sources which are low activity and exempt from licensing. It also contains an 11-piece absorber set which is stored within the instrument cabinet. A student experiment manual and teacher's guide covering a wide range of laboratory exercises, a serial cable and software is included. The bidirectional interface features counter control from the computer and data collection in computer files.
Essential Nuclear Lab Systems
Includes the absorbers and software, but, does not include sources, lab manuals, or serial cable. This all-inclusive Nuclear Lab Station comes with three radiation sources, an 11-piece absorber set a student experiment manual and guide, computer software and cable.
Description
Designed for high school and junior college level study. Experiments include range of alpha particles, absorption of beta particles, inverse square law, half-life decay, detection efficiency counting statistics, plus much more!
Flexible yet simple
- Minimum control ensures easy operation
- Digitally-controlled variable voltage setting allow students to plot the Geiger Plateau
- By connecting to an IBM, students can run experiments from a laboratory work station and display data directly on the computer screen
- Software stores each measurement in a data file
- Data file can be transferred to a spreadsheet for graphing and presentation
- Individual sealed sources
- Uncalibrated and exempt from NRC and state licensing
- Use them to replace older decayed sources or expand your inventory
- General purpose uncalibrated check sources
- Supplied as 1 in. diameter solid discs packages in plastic containers
Specifications
1 in. | |
Basic nuclear demonstration | |
Disc source, Po-210 alpha |
2.5cm | |
The nuclear lab station, essential nuclear lab systems |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title