Fisherbrand™ Basic Protection Disposable Polypropylene Medical Blue Lab Coats
Single-use lab coats provide economical, lightweight and breathable protection
 View more versions of this product
Catalog No. 17-100-891A
  Check Availability
Add to Cart
    Specifications
    Length (English) 39 in.
    No. of Pockets 3
    Latex Free Yes
    Material Polypropylene
    Size Large
    Disposable Yes
    Collar Type Standard
    Closure Type Snap
    Product Type Basic protection; Three pockets
    Color Blue
    Cuff Style Knit
  • Description & Specifications
Related Products
Product Content Feedback