Fisherbrand™ Basic Protection Disposable Polypropylene Navy Lab Coats
Provide economical, lightweight, and breathable protection. Fisherbrand™ Basic Protection Disposable Polypropylene Navy Lab Coats are made of spunbond polypropylene and feature knit cuffs and snap closures.
 View more versions of this product
Catalog No. 17-100-947
  Check Availability
Add to Cart
    Specifications
    Material Polypropylene
    Size Medium
    Latex Free Yes
    Product Type Basic protection; Three pockets
    Length (English) 38 in.
    No. of Pockets 3
    Disposable Yes
    Collar Type Standard
    Closure Type Snap
    Color Navy
    Cuff Style Knit
    • Roomy pockets for securely storing items like pens or small instruments
    • Latex-free spunbond polypropylene construction
    • High level of breathability
    • Knit cuffs hold sleeves in place
    • Three pockets and a five-snap closure
    • Standard collar
    • Disposable
    • Fabric weight: 48g/m
    • 10/Pk; 30/Cs
    • Also available in white
    • Medium
    Note: Other materials, colors, and options are available. See Fisherbrand Basic Protection Disposable Polypropylene Lab Coats with Elastc Cuffs, and Fisherbrand Basic Protection Disposable SMS Lab Coats in a variety of colors.
  • Description & Specifications
Related Products
Product Content Feedback