Learn More
Fisherbrand™ Basic Steel Work Bench Complete Plumbing Assembly
Fisherbrand™ Plumbing Assemblies are to be used with Fisherbrand Basic Work Bench.
$439.00
Specifications
Includes Male Quick Connect, System Hose and Service Turret
Material Reinforced PVC Hose, Polished Chrome Turret
View More Specs
Products 3
Catalog Number Color Type Price Quantity & Availability  
Catalog Number Color Type Price Quantity & Availability  
06-000-677
View Documents
Color Coded Orange Air Service Complete Plumbing Assembly Each for $439.00
Add to cart
06-000-678
View Documents
Color Coded Yellow Nitrogen Service Complete Plumbing Assembly Each for $439.00
Add to cart
06-000-679
View Documents
Color Coded Brown Vacuum Service Complete Plumbing Assembly Each for $439.00
Add to cart
Description
Description
  • Plumbing assemblies include a fine control needle valve service turret with shank, a service hose, and a male quick connect fitting.
  • These complete assemblies are designed specifically for use with Basic Work Bench in a laboratory environment.
  • All quick connects have 316 stainless steel internal components.
  • Keyed quick connect members will only couple with components of the same key which are identified by a color code. 
Specifications
Specifications
Male Quick Connect, System Hose and Service Turret
Reinforced PVC Hose, Polished Chrome Turret