Learn More
Basix Disposable Borosilicate Glass Culture Tubes Available on GSA/VA Contract for Federal Government customers only.
Disposable glass culture tubes for a variety of tissue culture, cell washing, and clinical chemistry applications.
 View more versions of this product
Catalog No. 14-955-413
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • 70 expansion borosilicate glass
  • Tubes are packed in convenient trays for ease of use on bench tops and in drawers
  • Sturdy, uniform bottom, and consistent length
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
100 mm
Round Bottom
16 mm
Borosilicate Glass
15 mL
100/Cs.
Disposable Culture Tube
Clear