Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bassoon Mouse anti-Human, Mouse, Rat, Bovine, Canine, Clone: SAP7F407, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289079

Catalog No. NB036664

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »


Description

Description

Bassoon Monoclonal antibody specifically detects Bassoon in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications

Specifications

Bassoon
SAP7F407
PBS, pH 7.2, containing 50% glycerol
bassoon (presynaptic cytomatrix protein)
Recombinant rat Bassoon.
Protein G purified
RUO
Primary
8927
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Purified
Mouse
IgG2a Kappa
50 μg
Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat, Bovine, Canine
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit