Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bat Immunoglobulin Mouse anti-Bat, Alexa Fluor™ 350, Clone: BT1-4F10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP223483AF350

Catalog No. NP223483A30


Add to cart

Description

Description

Bat Immunoglobulin Monoclonal antibody specifically detects Bat Immunoglobulin in Bat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Bat Immunoglobulin
BT1-4F10
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
Microchiropteran bat (Eptesicus fuscus) spleen cells.
Protein G purified
Store at 4°C in the dark.
Monoclonal
Bat
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG1
0.1 mL
Primary
Immunoglobulin L (Light) chain. This antibody reacts with Microchiropteran bats (confirmed against big brown, little brown, Red, Silverhair, Hoary bats), also reacts with porcine. This antibody does not react with Megachiropteran bats (confirmed against P. poliocephalus, P. giganteus, P. rodricensis, R. aegyptiacus, P. vampyrus, E. helvum, P.pumilus, P. hypomelanus), human, mouse, rat, hamster, canine, feline, guinea pig, and bovine.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit