Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPANK1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549632

Catalog No. PA549632

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene is located in a cluster of HLA-B-associated transcripts, which is included in the human major histocompatability complex III region. This gene encodes a protein which is thought to play a role in immunity. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq, Nov 2010]
Specifications

Specifications

GPANK1
Polyclonal
PBS with 0.09% sodium azide
O95872, Q61858
G patch domain and ankyrin repeat-containing protein 1, Ankyrin repeat domain-containing protein 59, G patch domain-containing protein 10, HLA-B-associated transcript 4, Protein G5, GPANK1, ANKRD59, BAT4, G5, GPATCH10
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7918, 81845
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GPANK1
Liquid
GPANK1
KLH conjugated synthetic peptide between 275-302 amino acids from the C-terminal region of human BAT4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit