Learn More
Aviva Systems Biology Corp BAT5 Antibody - middle region (ARP49633_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49633_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BAT5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BAT5. Alternate names: D6S82E, NG26, BAT5, PP199ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60131