Bata Viking Galoshes and Overboots

Manufacturer: Onguard Industries LLCSupplier Diversity Partner 860102X00

 View more versions of this product

Catalog No. 17-986-363

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Used in construction, farming, utilities and food processing
    Gender Male
    Height (English) 4 in.
    Lug Type Kick off
    Sole Features 4-way cleated, slip resistant outsole
    Waterproof Yes
    Color Black
    For Use With (Equipment) 14 to 15 size
    Toe Style Plain
    Material Flexothane, PVC
    Size 2X-Large

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit