Learn More
BATF Mouse anti-Human, Alexa Fluor 350, Clone: 687706, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8054U100UG
Catalog No. FAB8054U100
Add to cart
This item is not returnable. View return policy
Description
Description
BATF Monoclonal antibody specifically detects BATF in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
Specifications
BATF
Monoclonal
0.2 mg/mL
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Q16520
basic leucine zipper transcription factor, ATF-like, BATF1basic leucine zipper transcriptional factor ATF-like, B-ATFSFA2, B-cell-activating transcription factor, SFA2, SFA-2activating transcription factor B, SF-HT-activated gene 2 protein
Mouse
IgG2b
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human BATF in ELISA and Western Blot.
Flow Cytometry
687706
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Purified
BATF
E. coli-derived recombinant human BATF Met1-Pro125 Accession # Q16520
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10538
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)