Learn More
Human BATF Alexa Fluor 488-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB8054G100UG
Catalog No. FAB8054G100
Add to Cart
This item is not returnable. View return policy
Description
Description
BATF Monoclonal specifically detects BATF in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
Specifications
BATF
Monoclonal
0.2 mg/mL
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Q16520
BATF
E. coli-derived recombinant human BATF Met1-Pro125 Accession # Q16520
100 μg
Primary
Detects human BATF in ELISA and Western Blot.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2b
Flow Cytometry
687706
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
basic leucine zipper transcription factor, ATF-like, BATF1basic leucine zipper transcriptional factor ATF-like, B-ATFSFA2, B-cell-activating transcription factor, SFA2, SFA-2activating transcription factor B, SF-HT-activated gene 2 protein
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
10538
Human
Purified
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)