Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BATF Mouse anti-Human, Alexa Fluor 594, Clone: 687706, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8054T100UG
This item is not returnable.
View return policy
Description
BATF Monoclonal specifically detects BATF in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
BATF | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q16520 | |
BATF | |
E. coli-derived recombinant human BATF Met1-Pro125 Accession # Q16520 | |
100 μg | |
Primary | |
Detects human BATF in ELISA and Western Blot. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Flow Cytometry | |
687706 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
basic leucine zipper transcription factor, ATF-like, BATF1basic leucine zipper transcriptional factor ATF-like, B-ATFSFA2, B-cell-activating transcription factor, SFA2, SFA-2activating transcription factor B, SF-HT-activated gene 2 protein | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10538 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)