Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BATF3 Mouse anti-Human, Alexa Fluor 700, Clone: 841702, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7437N100UG
This item is not returnable.
View return policy
Description
BATF3 Monoclonal specifically detects BATF3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
BATF3 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9NR55 | |
BATF3 | |
E. coli-derived recombinant human BATF3 Met1-Arg127 Accession # Q9NR55 | |
100 μg | |
Primary | |
Detects human BATF3 in ELISA. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Flow Cytometry | |
841702 | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
basic leucine zipper transcription factor, ATF-like 3, basic leucine zipper transcriptional factor ATF-like 3, B-ATF-3, FLJ36352, FLJ37535,21 kDa small nuclear factor isolated from T-cells, JDP1, Jun dimerization protein 1, Jun dimerization protein p21SNFT, JUNDM1, SNFT, SNFT21-kD small nuclear factor isolated from T cells | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
55509 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)