Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bathophenanthrolinedisulfonic acid disodium salt hydrate, 98%

Click to view available options
Quantity:
250mg
1g
5g
52746-49-3
C24H16N2Na2O7S2
554.499
MFCD00149310
PFNPMHXHQCVXED-UHFFFAOYSA-L
4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid disodium salt hydrate
54669730
disodium;4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzenesulfonate;hydrate
C1=CC(=CC=C1C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-].O.[Na+].[Na+]
52746-49-3
C24H16N2Na2O7S2
554.499
MFCD00149310
PFNPMHXHQCVXED-UHFFFAOYSA-L
4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid disodium salt hydrate
54669730
disodium;4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzenesulfonate;hydrate
C1=CC(=CC=C1C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-].O.[Na+].[Na+]
Applications
Bathophenanthrolinedisulfonic acid disodium salt hydrate is used as colorimetric reagent for detection of iron. It is widely used for aqueous iron determination without the usage of solvent for extraction. It is used as precursor in the preparation of anionic Europium(III) disulfonated bathophenanthroline tris (dibenzoylmethanate) complex and ruthenium II tris (bathophenanthroline disulfonate). It plays a vital role in the catyltic oxidation of 2-hexanol by making it a water soluble complex with palladium.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Melting Point >300°C
Quantity 1g
Formula Weight 536.48 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Bathophenanthrolinedisulfonic acid disodium salt hydrate

RTECSNumber : SF8433000

TSCA : TSCA