Alfa Aesar™ Bathophenanthrolinedisulfonic acid disodium salt hydrate, 98%

Manufacturer: Alfa Aesar™ B23244MD

Quantity 250mg

 View more versions of this product

Catalog No. AAB23244MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 52746-49-3
    Molecular Formula C24H14N2Na2O6S2·xH2O
    Formula Weight 536.48 (Anhydrous)
    MDL Number MFCD00149310
    Synonym 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid disodium salt hydrate
    Chemical Name or Material Bathophenanthrolinedisulfonic acid disodium salt hydrate

    Specifications

    Formula Weight 536.48 (Anhydrous)
    TSCA TSCA
    Molecular Formula C24H14N2Na2O6S2·xH2O
    MDL Number MFCD00149310
    CAS 52746-49-3
    Synonym 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid disodium salt hydrate
    Melting Point >300°C
    Quantity 250mg
    RTECS Number SF8433000
    Chemical Name or Material Bathophenanthrolinedisulfonic acid disodium salt hydrate
    Assay 98%

    Description

    Applications
    Bathophenanthrolinedisulfonic acid disodium salt hydrate is used as colorimetric reagent for detection of iron. It is widely used for aqueous iron determination without the usage of solvent for extraction. It is used as precursor in the preparation of anionic Europium(III) disulfonated bathophenanthroline tris (dibenzoylmethanate) complex and ruthenium II tris (bathophenanthroline disulfonate). It plays a vital role in the catyltic oxidation of 2-hexanol by making it a water soluble complex with palladium.

    Solubility
    Soluble in water.

    Notes
    Incompatible with strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit